2-Chloro-5-(trifluoromethyl)benzaldehyde ≥98%

Supplier: Matrix Scientific

002675-5G 002675-25G
101784-408EA 23.08 USD
101784-408 101784-410
2-Chloro-5-(trifluoromethyl)benzaldehyde ≥98%
2-Chloro-5-(trifluoromethyl)benzaldehyde

Hazard code(s): Irritant

Formula: C₈H₄ClF₃O
MW: 208.57 g/mol
Boiling Pt: 42…44 °C ( 1.5mm)
Density: 1.435 g/cm³
MDL Number: MFCD00134430
CAS Number: 82386-89-8

Order Now

Specification Test Results

Boiling point 42-44°C/ 1.
Density 1.435
ND20 1.488

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR