(2-Chloro-5-(trifluoromethyl)phenyl)acetic acid ≥95%

Supplier: Matrix Scientific
Warning

Synonyms: 2-(2-Chloro-5-(trifluoromethyl)phenyl)acetic acid

002651-1G 002651-5G
101784-396EA 76.92 USD
101784-396 101784-398
(2-Chloro-5-(trifluoromethyl)phenyl)acetic acid ≥95%
(2-Chloro-5-(trifluoromethyl)phenyl)acetic acid

Hazard code(s): Irritant

Formula: C₉H₆ClF₃O₂
MW: 238.59 g/mol
Melting Pt: 112…114 °C
MDL Number: MFCD01631471
CAS Number: 22893-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR