2-Chloro-5-(trifluoromethyl)benzoic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-trifluoromethylbenzoic acid, 6-Chloro-α,α,α-trifluoro-m-toluic acid

002641-5G 002641-25G
101784-370EA 35.38 USD
101784-370 101784-372
2-Chloro-5-(trifluoromethyl)benzoic acid ≥98%
2-Chloro-5-(trifluoromethyl)benzoic acid

Hazard code(s): Irritant

Formula: C₈H₄ClF₃O₂
MW: 224.57 g/mol
Melting Pt: 93…96 °C
MDL Number: MFCD00061094
CAS Number: 657-06-7

Order Now

Specification Test Results

Melting point 93-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR