(3-Fluoro-5-(trifluoromethyl)phenyl)acetic acid ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 3-Fluoro-5-(trifluoromethyl)phenylacetic Acid

002637-1G 002637-5G 002637-25G
101784-360EA 30.77 USD
101784-360 101784-362 101784-364
(3-Fluoro-5-(trifluoromethyl)phenyl)acetic acid ≥98%
(3-Fluoro-5-(trifluoromethyl)phenyl)acetic acid

Hazard code(s): Irritant

Formula: C₉H₆F₄O₂
MW: 222.14 g/mol
MDL Number: MFCD00061188
CAS Number: 195447-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR