3-Fluoro-5-(trifluoromethyl)benzylamine ≥97%

Supplier: Matrix Scientific

Synonyms: (3-Fluoro-5-(trifluoromethyl)phenyl)methanamine

002631-1G 002631-5G
101784-352EA 93.85 USD
101784-352 101784-354
3-Fluoro-5-(trifluoromethyl)benzylamine ≥97%
3-Fluoro-5-(trifluoromethyl)benzylamine

Hazard code(s): Corrosive, Irritant

Formula: C₈H₇F₄N
MW: 193.14 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 46 °C
Density: 1.319 g/cm³ (20 °C)
MDL Number: MFCD00061163
CAS Number: 150517-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR