2-Fluoro-5-(trifluoromethyl)benzylamine ≥97% for synthesis

Supplier: Matrix Scientific

002630-1G 002630-5G
101784-348EA 41.54 USD
101784-348 101784-350
2-Fluoro-5-(trifluoromethyl)benzylamine ≥97% for synthesis
2-Fluoro-5-(trifluoromethyl)benzylamine

Hazard code(s): Irritant

Formula: C₈H₇F₄N
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 46 °C
MDL Number: MFCD00061162
CAS Number: 199296-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR