2-Fluoro-5-(trifluoromethyl)benzyl alcohol ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: (2-Fluoro-5-(trifluoromethyl)phenyl)methanol

002628-1G 002628-5G 002628-25G
101784-338EA 30.77 USD
101784-338 101784-340 101784-342
2-Fluoro-5-(trifluoromethyl)benzyl alcohol ≥97% for synthesis
2-Fluoro-5-(trifluoromethyl)benzyl alcohol

Hazard code(s): Irritant

Formula: C₈H₆F₄O
MDL Number: MFCD00061167
CAS Number: 207974-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR