3-Fluoro-5-(trifluoromethyl)benzamide ≥97%

Supplier: Matrix Scientific

002625-1G 002625-5G
101784-326EA 36.92 USD
101784-326 101784-328
3-Fluoro-5-(trifluoromethyl)benzamide ≥97%
3-Fluoro-5-(trifluoromethyl)benzamide

Hazard code(s): Irritant

Formula: C₈H₅F₄NO
MDL Number: MFCD00061149
CAS Number: 207986-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR