2-Fluoro-5-(trifluoromethyl)benzamide ≥97%

Supplier: Matrix Scientific

002624-1G 002624-5G
101784-322EA 29.23 USD
101784-322 101784-324
2-Fluoro-5-(trifluoromethyl)benzamide ≥97%
2-Fluoro-5-(trifluoromethyl)benzamide

Hazard code(s): Irritant

Formula: C₈H₅F₄NO
MDL Number: MFCD00061148
CAS Number: 207919-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR