3-Fluoro-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: Matrix Scientific

002623-1G 002623-5G 002623-25G
101784-316EA 20 USD
101784-316 101784-318 101784-320
3-Fluoro-5-(trifluoromethyl)benzaldehyde ≥97%
3-Fluoro-5-(trifluoromethyl)benzaldehyde

Hazard code(s): Irritant

Formula: C₈H₄F₄O
Boiling Pt: 175 °C
Density: 1.376 g/cm³
MDL Number: MFCD00061312
CAS Number: 188815-30-7

Order Now

Specification Test Results

Boiling point 175°C
Density 1.376
ND20 1.45


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR