α,α,α,5-Tetrafluoro-m-toluic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-5-trifluoromethylbenzoic acid, 3-Fluoro-5-(trifluoromethyl)benzoic acid

002616-5G 002616-25G
101784-298EA 26.15 USD
101784-298 101784-300
α,α,α,5-Tetrafluoro-m-toluic acid ≥97%
α,α,α,5-Tetrafluoro-m-toluic acid

Hazard code(s): Irritant

Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
Melting Pt: 104…108 °C
MDL Number: MFCD00061293
CAS Number: 161622-05-5

Order Now

Specification Test Results

Melting point 104-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR