α,α,α,6-Tetrafluoro-m-toluic acid ≥96%

Supplier: Matrix Scientific

002614-5G 002614-25G 002614-100G
101784-292EA 21.54 USD
101784-292 101784-294 101784-296
α,α,α,6-Tetrafluoro-m-toluic acid ≥96%
α,α,α,6-Tetrafluoro-m-toluic acid

Hazard code(s): Irritant

Formula: C₈H₄F₄O₂
Melting Pt: 101…102 °C
MDL Number: MFCD00061292
CAS Number: 115029-23-7

Order Now

Specification Test Results

Melting point 101-102°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR