3-Fluoro-4-nitrotoluene ≥98%
Supplier: Matrix Scientific
002599-25G
002599-100G
101784-260EA
52.31
USD
101784-260
101784-262
3-Fluoro-4-nitrotoluene ≥98%
3-Fluoro-4-nitrotoluene
Hazard code(s): Irritant
Formula:
C₇H₆FNO₂ MW: 155.13 g/mol Boiling Pt: 249 °C (1013 hPa) Melting Pt: 53 °C Density: 1.438 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007053 CAS Number: 446-34-4 EINECS: 207-166-5 UN: 2811 ADR: 6.1,III |
Specification Test Results
Boiling point | 97-98°C/3mm |
Melting point | 53-55°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...