2-Fluoro-4-nitrophenol ≥99%
Supplier: Matrix Scientific
002596-25G
002596-100G
101784-248EA
33.85
USD
101784-248
101784-250
2-Fluoro-4-nitrophenol ≥99%
2-Fluoro-4-nitrophenol
Hazard code(s): Irritant
Formula:
C₆H₄FNO₃ Melting Pt: 120…122 °C |
MDL Number:
MFCD00051970 CAS Number: 403-19-0 |
Specification Test Results
Melting point | 120-122°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...