3-Chloro-4-nitrobenzotrifluoride ≥97% for synthesis
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
1-Chloro-2-nitro-5-(trifluoromethyl)benzene, 3-Chloro-4-nitrobenzotrifluoride, 2-Chloro-1-nitro-4-(trifluoromethyl)benzene
002593-1G
002593-5G
101784-240EA
44.62
USD
101784-240
101784-242
3-Chloro-4-nitrobenzotrifluoride ≥97% for synthesis
3-Chloro-4-nitrobenzotrifluoride
Hazard code(s): Irritant, Irritant-Harmful
Formula:
C₇H₃ClF₃NO₂ Boiling Pt: 54 °C (0.4mm) |
MDL Number:
MFCD00272564 CAS Number: 402-11-9 EINECS: 206-936-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...