2-Chloro-4-nitrobenzhydrazide for synthesis

Supplier: Matrix Scientific

002592-5G 002592-25G
101784-236EA 123.08 USD
101784-236 101784-238
2-Chloro-4-nitrobenzhydrazide for synthesis
2-Chloro-4-nitrobenzhydrazide

Hazard code(s): Irritant

Formula: C₇H₆ClN₃O₃
Melting Pt: 159…162 °C
MDL Number: MFCD00017047
CAS Number: 67345-78-2

Order Now

Specification Test Results

Melting point 159-162°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR