2-Fluoro-4-nitrobenzoic acid ≥95%

Supplier: Matrix Scientific
Warning

002591-5G 002591-25G
101784-232EA 41.54 USD
101784-232 101784-234
2-Fluoro-4-nitrobenzoic acid ≥95%
2-Fluoro-4-nitrobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Melting Pt: 174…177 °C
MDL Number: MFCD00275565
CAS Number: 403-24-7

Order Now

Specification Test Results

Melting point 174-177°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR