2-Fluoro-4-nitroanisole ≥98% for synthesis

Supplier: Matrix Scientific
Warning

002589-5G 002589-25G
101784-224EA 47.69 USD
101784-224 101784-226
2-Fluoro-4-nitroanisole ≥98% for synthesis
2-Fluoro-4-nitroanisole

Hazard code(s): Irritant

Formula: C₇H₆FNO₃
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00061095
CAS Number: 455-93-6

Order Now

Specification Test Results

Melting point 103-105°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR