2-Fluoro-4-methylpyridine ≥98%

Supplier: Matrix Scientific
Danger

Synonyms: 2-Fluoro-4-picoline

002586-25G 002586-100G
101784-218EA 24.62 USD
101784-218 101784-220
2-Fluoro-4-methylpyridine ≥98%
2-Fluoro-4-methylpyridine

Hazard code(s): Irritant

Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 157 °C (1013 hPa)
Density: 1.0805 g/cm³ (25 °C)
MDL Number: MFCD00041224
CAS Number: 461-87-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 155-156°C
ND20 1.4725

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR