2-Chloro-p-cresol ≥95%

Supplier: Matrix Scientific
Warning

002584-5G 002584-25G
101784-214EA 32.31 USD
101784-214 101784-216
2-Chloro-p-cresol ≥95%
2-Chloro-p-cresol

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₇H₇ClO
Boiling Pt: 195…197 °C
Melting Pt: 76 °C
Density: 1.21 g/cm³
MDL Number: MFCD00070639
CAS Number: 6640-27-3
EINECS: 229-656-8
UN: 2669
ADR: 6.1,II

Order Now

Specification Test Results

Boiling point 195-197°C
Density 1.21


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR