3-Chloro-p-toluic acid ≥98%

Supplier: Matrix Scientific
Warning

002579-5G 002579-25G
101784-198EA 21.54 USD
101784-198 101784-200
3-Chloro-p-toluic acid ≥98%
3-Chloro-p-toluic acid

Hazard code(s): Irritant

Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Melting Pt: 203…204 °C
MDL Number: MFCD00045841
CAS Number: 5162-82-3

Order Now

Specification Test Results

Melting point 203-204°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR