3-Fluoro-4-methylbenzamide

Supplier: Matrix Scientific

002574-1G 002574-5G
101784-190EA 36.92 USD
101784-190 101784-192
3-Fluoro-4-methylbenzamide
3-Fluoro-4-methylbenzamide
Formula: C₈H₈FNO
MDL Number: MFCD01631412
CAS Number: 170726-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR