3-Fluoro-4-methylbenzaldehyde ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Fluoro-4-tolualdehyde, 3-Fluoro-p-tolylaldehyde, 3-Fluoro-p-tolualdehyde
002573-5G
002573-25G
101784-186EA
38.46
USD
101784-186
101784-188
3-Fluoro-4-methylbenzaldehyde ≥97%
3-Fluoro-4-methylbenzaldehyde
Hazard code(s): Irritant
Formula:
C₈H₇FO MW: 138.14 g/mol |
MDL Number:
MFCD01631532 CAS Number: 177756-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...