3-Fluoro-4-methoxybenzoic acid

Supplier: Matrix Scientific

002555-25G 002555-100G
101784-160EA 67.69 USD
101784-160 101784-162
3-Fluoro-4-methoxybenzoic acid
3-Fluoro-4-methoxybenzoic acid

Hazard code(s): Irritant

Formula: C₈H₇FO₃
MW: 170.14 g/mol
Melting Pt: 211…213 °C
MDL Number: MFCD00060675
CAS Number: 403-20-3

Order Now

Specification Test Results

Melting point 211-213°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR