3-Chloro-4-iodotoluene ≥99%

Supplier: Matrix Scientific

002553-25G 002553-100G
101784-150EA 95.38 USD
101784-150 101784-152
3-Chloro-4-iodotoluene ≥99%
3-Chloro-4-iodotoluene

Hazard code(s): Irritant

Formula: C₇H₆ClI
MW: 252.48 g/mol
Boiling Pt: 140 °C (22mm)
Density: 1.82 g/cm³
MDL Number: MFCD00079734
CAS Number: 116632-42-9

Order Now

Specification Test Results

Boiling point 140°C/22mm
Density 1.82

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR