2-Chloro-1-iodo-4-nitrobenzene ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-4-iodonitrobenzene

002550-5G 002550-25G 002550-100G
101784-142EA 36.92 USD
101784-142 101784-144 101784-146
2-Chloro-1-iodo-4-nitrobenzene ≥98%
2-Chloro-1-iodo-4-nitrobenzene

Hazard code(s): Irritant, Light Sensitive

Formula: C₆H₃ClINO₂
MW: 283.45 g/mol
Melting Pt: 103 °C
MDL Number: MFCD00024589
CAS Number: 41252-96-4
EINECS: 255-278-8

Order Now

Specification Test Results

Melting point 94-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR