3-Chloro-4-iodobenzotrifluoride ≥97%

Supplier: Matrix Scientific

002549-1G 002549-5G 002549-25G
101784-136EA 18.46 USD
101784-136 101784-138 101784-140
3-Chloro-4-iodobenzotrifluoride ≥97%
3-Chloro-4-iodobenzotrifluoride

Hazard code(s): Irritant

Formula: C₇H₃ClF₃I
Boiling Pt: 58…60 °C (0.2mm)
MDL Number: MFCD00042562
CAS Number: 141738-80-9

Order Now

Specification Test Results

Boiling point 58-60°C/0.2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR