2-Fluoro-4-iodoaniline ≥98%

Supplier: Matrix Scientific

002545-25G 002545-100G
101784-128EA 38.46 USD
101784-128 101784-130
2-Fluoro-4-iodoaniline ≥98%
2-Fluoro-4-iodoaniline

Hazard code(s): Irritant-Harmful, Light Sensitive

Formula: C₆H₅FIN
MW: 237.02 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 55…57 °C
MDL Number: MFCD00011738
CAS Number: 29632-74-4

Order Now

Specification Test Results

Melting point 55-57°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR