3-Fluoro-4-hydroxybenzoic acid ≥95%
Supplier: Matrix Scientific
002535-5G
002535-25G
101784-084EA
56.92
USD
101784-084
101784-086
3-Fluoro-4-hydroxybenzoic acid ≥95%
3-Fluoro-4-hydroxybenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₅FO₃ MW: 156.11 g/mol Melting Pt: 157…159 °C |
MDL Number:
MFCD00143161 CAS Number: 350-29-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...