2-Fluoro-4-hydroxybenzonitrile ≥99% for synthesis

Supplier: Matrix Scientific

002534-5G 002534-25G 002534-100G
101784-078EA 12.31 USD
101784-078 101784-080 101784-082
2-Fluoro-4-hydroxybenzonitrile ≥99% for synthesis
2-Fluoro-4-hydroxybenzonitrile

Hazard code(s): Irritant

Formula: C₇H₄FNO
Boiling Pt: 261 °C (1013 hPa)
Melting Pt: 123…125 °C
Density: 1.45 g/cm³ (20 °C)
MDL Number: MFCD00192177
CAS Number: 82380-18-5
EINECS: 422-810-7

Order Now

Specification Test Results

Melting point 123-125°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR