5-Chloro-2-(trifluoromethyl)phenol ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Chloro-4-(trifuloromethyl)phenol, 1-Chlor-2-hydroxy-5-(trifluormethyl)benzol, 3-Chloro-4-hydroxybenzotrifluoride, 2-Chloro-4-(trifluoromethyl)phenol
002533-5G
002533-25G
101784-074EA
21.54
USD
101784-074
101784-076
5-Chloro-2-(trifluoromethyl)phenol ≥97%
5-Chloro-2-(trifluoromethyl)phenol
Hazard code(s): Irritant
Formula:
C₇H₄ClF₃O MW: 196.56 g/mol |
MDL Number:
MFCD00042217 CAS Number: 35852-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...