3-Chloro-4-fluorotoluene ≥98%

Supplier: Matrix Scientific
Danger

002526-5G 002526-25G
101784-058EA 21.54 USD
101784-058 101784-060
3-Chloro-4-fluorotoluene ≥98%
3-Chloro-4-fluorotoluene

Hazard code(s): Irritant

Formula: C₇H₆ClF
Boiling Pt: 167…169 °C
Density: 1.18 g/cm³
MDL Number: MFCD00079730
CAS Number: 1513-25-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 167-169°C
Density 1.18


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR