2-Chlor-4-fluorothiophenol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chlor-4-fluorobenzene-1-thiol, 2-Chloro-4-fluorobenzenethiol, 1-Chloro-3-fluoro-6-mercaptobenzene, 2-Chlor-4-fluorobenzenethiol

002524-1G 002524-5G
101784-054EA 30.77 USD
101784-054 101784-056
2-Chlor-4-fluorothiophenol ≥95%
2-Chlor-4-fluorothiophenol

Hazard code(s): Irritant, Stench

Formula: C₆H₄ClFS
MW: 162.61 g/mol
Boiling Pt: 86…88 °C (15mm)
MDL Number: MFCD00052936
CAS Number: 175277-99-3

Order Now

Specification Test Results

Boiling point 86-88°C/15m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR