3-Chlor-4-fluorothiophenol tech

Supplier: Matrix Scientific

Synonyms: 1-Chloro-2-fluoro-5-mercaptobenzene, 3-Chloro-4-fluorobenzenethiol, 3-Chlor-4-fluorobenzenethiol, 3-Chlor-4-fluorobenzene-1-thiol

002523-1G 002523-5G
101784-050EA 60 USD
101784-050 101784-052
3-Chlor-4-fluorothiophenol tech
3-Chlor-4-fluorothiophenol

Hazard code(s): Toxic, Stench

Formula: C₆H₄ClFS
MW: 162.61 g/mol
Boiling Pt: 68…70 °C (2mm)
MDL Number: MFCD00052938
CAS Number: 60811-23-6

Order Now

Specification Test Results

Boiling point 68-70°C/2mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR