(3-Chloro-4-fluorophenyl)acetic acid ≥97%

Supplier: Matrix Scientific
Warning

002518-5G 002518-25G
101784-040EA 35.38 USD
101784-040 101784-042
(3-Chloro-4-fluorophenyl)acetic acid ≥97%
(3-Chloro-4-fluorophenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₃ClF)CH₂COOH
MDL Number: MFCD01631555
CAS Number: 705-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR