2-Chloro-1-fluoro-4-nitrobenzene ≥98%
Supplier: Matrix Scientific
Synonyms:
3-Chloro-4-fluoronitrobenzene, 1-Chloro-2-fluoro-5-nitrobenzene
002515-25G
002515-100G
101784-026EA
24.62
USD
101784-026
101784-028
2-Chloro-1-fluoro-4-nitrobenzene ≥98%
2-Chloro-1-fluoro-4-nitrobenzene
Hazard code(s): Irritant
Formula:
C₆H₃ClFNO₂ MW: 175.55 g/mol Boiling Pt: 227…232 °C Melting Pt: 44…47 °C Density: 1.61 g/cm³ |
MDL Number:
MFCD00007206 CAS Number: 350-30-1 EINECS: 206-499-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Melting point | 44-47°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...