2-Chloro-4-fluorobenzonitrile ≥99%

Supplier: Matrix Scientific

002497-25G 002497-100G
101784-002EA 43.08 USD
101784-002 101784-004
2-Chloro-4-fluorobenzonitrile ≥99%
2-Chloro-4-fluorobenzonitrile

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₇H₃ClFN
MW: 155.56 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 63 - 65 °C
MDL Number: MFCD00042523
CAS Number: 60702-69-4
EINECS: 262-384-8
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 63-65°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR