2-Chloro-4-fluorobenzoic acid ≥98%

Supplier: Matrix Scientific

002496-25G 002496-100G
101783-998EA 20 USD
101783-998 101784-000
2-Chloro-4-fluorobenzoic acid ≥98%
2-Chloro-4-fluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
Melting Pt: 187…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00010615
CAS Number: 2252-51-9
EINECS: 218-845-0

Order Now

Specification Test Results

Melting point 187-189°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR