3-Chloro-4-fluorobenzamide ≥97%

Supplier: Matrix Scientific

002495-1G 002495-5G
101783-994EA 21.54 USD
101783-994 101783-996
3-Chloro-4-fluorobenzamide ≥97%
3-Chloro-4-fluorobenzamide

Hazard code(s): Irritant

Formula: C₇H₅ClFNO
Melting Pt: 145…147 °C
MDL Number: MFCD00153092
CAS Number: 701-43-9

Order Now

Specification Test Results

Melting point 145-147°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR