2-Chloro-4-fluorobenzaldehyde ≥97%

Supplier: Matrix Scientific
Warning

002493-25G 002493-100G
101783-986EA 27.69 USD
101783-986 101783-988
2-Chloro-4-fluorobenzaldehyde ≥97%
2-Chloro-4-fluorobenzaldehyde

Hazard code(s): Irritant

Formula: C₇H₄ClFO
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 60 °C
MDL Number: MFCD00042527
CAS Number: 84194-36-5

Order Now

Specification Test Results

Melting point 60-63°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR