3-Chloro-4-fluorobenzoic acid ≥98%

Supplier: Matrix Scientific

002492-5G 002492-25G 002492-100G
101783-980EA 15.38 USD
101783-980 101783-982 101783-984
3-Chloro-4-fluorobenzoic acid ≥98%
3-Chloro-4-fluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄ClFO₂
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 136 °C
MDL Number: MFCD00042477
CAS Number: 403-16-7

Order Now

Specification Test Results

Melting point 133-135°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR