3-Chloro-4-fluoro-5-nitrobenzotrifluoride
Supplier: Matrix Scientific
Synonyms:
1-Chloro-2-fluoro-3-nitro-5-(trifluoromethyl)benzene, 3-Chloro-4-fluoro-5-nitrobenzotrifluoride
002488-1G
002488-5G
101783-972EA
60
USD
101783-972
101783-974
3-Chloro-4-fluoro-5-nitrobenzotrifluoride
3-Chloro-4-fluoro-5-nitrobenzotrifluoride
Hazard code(s): Irritant-Harmful
Formula:
C₇H₂ClF₄NO₂ MW: 243.54 g/mol Boiling Pt: 200 °C Density: 1.607 g/cm³ |
MDL Number:
MFCD00674108 CAS Number: 101646-02-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...