Methyl-3,4-difluorobenzoate
Supplier: Matrix Scientific
002484-25G
002484-100G
101783-964EA
20
USD
101783-964
101783-966
Methyl-3,4-difluorobenzoate
Methyl-3,4-difluorobenzoate
Hazard code(s): Irritant
Formula:
C₈H₆F₂O₂ Boiling Pt: 88 °C (15mm) |
MDL Number:
MFCD00276597 CAS Number: 369-25-5 |
Specification Test Results
Boiling point | 88°C/15mm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...