(3-Fluoro-4-(trifluoromethyl)phenyl)acetic acid ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 3-Fluoro-4-(trifluoromethyl)phenylacetic acid

002443-1G 002443-5G
101783-936EA 30.77 USD
101783-936 101783-938
(3-Fluoro-4-(trifluoromethyl)phenyl)acetic acid ≥98%
(3-Fluoro-4-(trifluoromethyl)phenyl)acetic acid

Hazard code(s): Irritant

Formula: C₉H₆F₄O₂
MW: 222.14 g/mol
MDL Number: MFCD00236303
CAS Number: 238754-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR