3-Fluoro-4-(trifluoromethyl)benzaldehyde ≥97%

Supplier: Matrix Scientific

002432-1G 002432-5G
101783-910EA 18.46 USD
101783-910 101783-912
3-Fluoro-4-(trifluoromethyl)benzaldehyde ≥97%
3-Fluoro-4-(trifluoromethyl)benzaldehyde

Hazard code(s): Irritant, Air Sensitive

Formula: C₈H₄F₄O
MDL Number: MFCD00236275
CAS Number: 204339-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR