2-Fluoro-4-(trifluoromethyl)benzyl alcohol ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: (2-Fluoro-4-(trifluoromethyl)phenyl)methanol

002426-1G 002426-5G
101783-902EA 29.23 USD
101783-902 101783-904
2-Fluoro-4-(trifluoromethyl)benzyl alcohol ≥97% for synthesis
2-Fluoro-4-(trifluoromethyl)benzyl alcohol

Hazard code(s): Irritant

Formula: C₈H₆F₄O
MDL Number: MFCD00070808
CAS Number: 197239-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR