2-Fluoro-4-(trifluoromethyl)benzamide ≥97%
Supplier: Matrix Scientific
002423-1G
002423-5G
101783-894EA
38.46
USD
101783-894
101783-896
2-Fluoro-4-(trifluoromethyl)benzamide ≥97%
2-Fluoro-4-(trifluoromethyl)benzamide
Hazard code(s): Irritant
Formula:
C₈H₅F₄NO Melting Pt: 140…142 °C |
MDL Number:
MFCD00061147 CAS Number: 207853-64-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...