2-Fluoro-4-(trifluoromethyl)benzaldehyde ≥97%
Supplier: Matrix Scientific
002422-5G
002422-25G
101783-890EA
56.92
USD
101783-890
101783-892
2-Fluoro-4-(trifluoromethyl)benzaldehyde ≥97%
2-Fluoro-4-(trifluoromethyl)benzaldehyde
Hazard code(s): Irritant, Store in Freezer
Formula:
C₈H₄F₄O MW: 192.11 g/mol Boiling Pt: 118 - 119 °C (1013 hPa) Density: 1.41 g/cm³ (20 °C) |
MDL Number:
MFCD00061310 CAS Number: 89763-93-9 UN: 1987 ADR: 3,III |
Specification Test Results
ND20 | 1.452 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...