α,α,α,2-Tetrafluoro-p-toluic acid ≥97%
Supplier: Matrix Scientific
002420-5G
002420-25G
101783-886EA
44.62
USD
101783-886
101783-888
α,α,α,2-Tetrafluoro-p-toluic acid ≥97%
α,α,α,2-Tetrafluoro-p-toluic acid
Hazard code(s): Irritant
Formula:
C₈H₄F₄O₂ Melting Pt: 160 °C |
MDL Number:
MFCD00010322 CAS Number: 115029-24-8 |
Specification Test Results
Melting point | 168-170°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...