3'-Bromo-4'-fluoroacetophenone ≥97%

Supplier: Matrix Scientific

002404-5G 002404-25G
101783-848EA 16.92 USD
101783-848 101783-850
3'-Bromo-4'-fluoroacetophenone ≥97%
3'-Bromo-4'-fluoroacetophenone

Hazard code(s): Irritant

Formula: C₈H₆BrFO
MW: 217.04 g/mol
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: 53…55 °C
MDL Number: MFCD00042466
CAS Number: 1007-15-4
EINECS: 213-750-0

Order Now

Specification Test Results

Melting point 53-55°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR